Learn More
Collagen I Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA127396
Catalog No. PA127396
Add to cart
Description
Description
PA1-27396 detects Collagen I from mouse samples. PA1-27396 will not react with human, rat, or chicken. PA1-27396 has been successfully used in radioimmune assay, ELISA, immunofluorescence and immunocytochemistry procedures. This product is unsuitable for western blot. The PA1-27396 immunogen is Collagen type I extracted and purified from mouse skin. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Collagen I is a Type I collagen with a triple helix structure comprised of two alpha-1 chains and one alpha-2 chain. Collagen I is a member of group I collagen (fibril-forming collagen) found in most connective tissues, and is abundant in bone, cornea, dermis and tendon. Mutations in the COL1A2 gene encoding the alpha-2 chain are associated with osteogenesis imperfecta types I-IV, Ehlers-Danlos syndrome type VIIB, recessive Ehlers-Danlos syndrome Classical type, idiopathic osteoporosis, and atypical Marfan syndrome. Symptoms associated with mutations in the COL1A2 gene which codes for the alpha-2 chain, however, tend to be less severe than mutations in the COL1A1 gene, reflecting the different role of alpha-2 chains in matrix integrity. Reciprocal translocations between chromosomes 17 and 22, where Collagen Type 1 genes and the gene for platelet derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified.
Specifications
Specifications
Collagen I
Polyclonal
Antibody with no preservative
P11087
Type I Collagen, Collagen 1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
12842
ELISA, Immunocytochemistry, Immunofluorescence, Radioimmune Assays
Unconjugated
Col1a1
Liquid
COL1A1
Collagen type I extracted and purified from mouse skin.
Chromatography
RUO
Antibody
Polyclonal
Mouse