Learn More
Collagen II Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136059
Catalog No. 01-672-242
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reconstitute with 1 mL of distilled water and add preservative if preferred.
COL2A1 is the alpha-1 chain of type II collagen, a fibrillar collagen found in cartilage and the vitreous humor of the eye. Mutations in this gene are associated with achondrogenesis, chondrodysplasia, early onset familial osteoarthritis, SED congenita, Langer-Saldino achondrogenesis, Kniest dysplasia, Stickler syndrome type I, and spondyloepimetaphyseal dysplasia Strudwick type. In addition, defects in processing chondrocalcin, a calcium binding protein that is the C-propeptide of this collagen molecule, are also associated with chondrodysplasia.
Specifications
Specifications
Collagen II
Polyclonal
whole serum with no preservative
P02458
COL2A1
Purified native collagen type II.
0.2mg
4° C
Primary
1280
ELISA, Immunohistochemistry (Frozen)
Unconjugated
COL2A1
Lyophilized
Rabbit
IgG
RUO
Antibody
Polyclonal
Human