Learn More
Collagen III Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136061
Catalog No. 01-672-244
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reconstitute with 1 mL of distilled water and add preservative if preferred.
This gene encodes a fibrillar collagen that is found in extensible connective tissues such as skin, lung, and the vascular system, frequently in association with type I collagen. Mutations in this gene are associated with Ehlers-Danlos syndrome type IV, and with aortic and arterial aneurysms. Although alternate transcripts have been detected for this gene, they are the result of mutations; these mutations alter splicing, often leading to the exclusion of multiple exons.
Specifications
Specifications
Collagen III
Polyclonal
whole serum with no preservative
P02461
EDS4A, FLJ34534
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
ELISA, Immunohistochemistry (Frozen)
Unconjugated
COL3A1
Lyophilized
COL3A1
Purified native collagen type III.
1 mL
4° C
Primary
1281