Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Collagen III Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA136062

Catalog No. 01-672-245



Description

Description

Reconstitute with 1 mL of distilled water and add preservative if preferred.

This gene encodes a fibrillar collagen that is found in extensible connective tissues such as skin, lung, and the vascular system, frequently in association with type I collagen. Mutations in this gene are associated with Ehlers-Danlos syndrome type IV, and with aortic and arterial aneurysms. Although alternate transcripts have been detected for this gene, they are the result of mutations; these mutations alter splicing, often leading to the exclusion of multiple exons.
Specifications

Specifications

Collagen III
Polyclonal
Unconjugated
COL3A1
Lyophilized
COL3A1
Purified collagen type III extracted from human skin.
Affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Immunohistochemistry (Frozen)
0.1 mg/mL
Antibody with no preservative
P02461
EDS4A, FLJ34534
Rabbit
IgG
1 mL
4° C
Primary
1281
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit