Learn More
Collagen III Rabbit anti-Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136146
Catalog No. 01-672-102
Add to cart
Description
Description
Reconstitute with 1 mL of distilled water and add preservative if preferred.
This gene encodes a fibrillar collagen that is found in extensible connective tissues such as skin, lung, and the vascular system, frequently in association with type I collagen. Mutations in this gene are associated with Ehlers-Danlos syndrome type IV, and with aortic and arterial aneurysms. Although alternate transcripts have been detected for this gene, they are the result of mutations; these mutations alter splicing, often leading to the exclusion of multiple exons.
Specifications
Specifications
Collagen III
Polyclonal
Antibody with no preservative
P13941
EDS4A, FLJ34534
Rabbit
IgG
100 μg
4° C
Primary
84032
ELISA, Immunohistochemistry (Frozen)
Unconjugated
Col3a1
Lyophilized
COL3A1
Purified native collagen type III extracted from rat tail tendon.
Affinity chromatography
RUO
Antibody
Polyclonal
Rat