Learn More
Collagen IV Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA128117
Catalog No. PA128117
Add to cart
Description
Description
PA1-28117 detects Collagen IV from mouse samples. PA1-28117 has been successfully used in immunofluorescence, immunohistochemistry (paraffin tissue), and ELISA applications. Positive control: skin, liver The PA1-28117 immunogen is: Recombit full length protein (Mouse). Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20° C or below, avoiding multiple freeze-thaw cycles.
Specifications
Specifications
Collagen IV
Polyclonal
Antibody with no preservative
P02463, P08122
Col4a1, Col4a2
Collagen Type IV extracted and purified from mouse tumor tissues.
Gravity Column Chromatography
RUO
Antibody
Polyclonal
Mouse
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Col4a2
Liquid
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
12826, 12827