Learn More
Collagen IV Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136063
Catalog No. 01-672-246
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reconstitute with 1 mL of distilled water and add preservative if preferred.
Collagen IV is a major constituent of the basement membranes along with laminins and enactins. It is composed of alpha 1 IV chain and alpha 2 IV chain in 2:1 ratio. It can form insoluble fibers with high tensile strength. Antibody to collagen IV is useful in detecting the loss of parts of basement membrane in carcinomas.
Specifications
Specifications
Collagen IV
Polyclonal
whole serum with no preservative
P02462, P08572
arresten
Rabbit
IgG
1 mL
4° C
Primary
1284, 1282
ELISA, Immunohistochemistry (Frozen)
Unconjugated
COL4A2
Lyophilized
COL4A1, COL4A2
Purified native collagen type IV.
Ammonium sulfate precipitation, Ion-exchange chromatography
RUO
Antibody
Polyclonal
Human