Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen IV Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA136065

Catalog No. 01-672-248


Add to cart

Description

Description

Reconstitute with 1 mL of distilled water and add preservative if preferred.

Collagen IV is a major constituent of the basement membranes along with laminins and enactins. It is composed of alpha 1 IV chain and alpha 2 IV chain in 2:1 ratio. It can form insoluble fibers with high tensile strength. Antibody to collagen IV is useful in detecting the loss of parts of basement membrane in carcinomas.
Specifications

Specifications

Collagen IV
Polyclonal
Unconjugated
COL4A2
Lyophilized
COL4A1, COL4A2
Purified collagen type IV extracted from human skin.
Affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Immunohistochemistry (Frozen)
0.1 mg/mL
Antibody with no preservative
P08572, P02462
arresten
Rabbit
IgG
1 mL
4° C
Primary
1282, 1284
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit