Learn More
MilliporeSigma™ anti-Collagen Type IV Ab-1; Polyclonal; ™ (Chemicon™) Non-distribution product as customer accommodation.
Manufacturer:  MilliporeSigma™ AB748/DEL
Catalog No. 50-174-045
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Antibody reacts with native and heat denatured human type IV collagen. Less than 10% cross reactivity with collagen types I, III, and V, and less than 1% with human collagen II. Antiserum was passed over immobilized human blood plasma proteins, and human collagen types I, III, and V. Human plasma proteins do not interfere with binding to collagen.
ELISA, Immunohistochemistry
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
Collagen Type IV Ab-1
Semi-purified
Purified human placental collagen type IV
RUO
Primary
Human
Unconjugated
Rabbit
50μg
Cell Structure
Polyclonal