Learn More
MilliporeSigma™ anti-Collagen Type IV Ab-3; Polyclonal; ™ (Chemicon™) Non-distribution product as customer accommodation.
Manufacturer:  MilliporeSigma™ AB769/DEL
Catalog No. 50-174-058
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Human and bovine type IV collagen as demonstrated by ELISA. Less than 10% cross reactivity with collagen types I, II, III, V, and VI. May show reactivity to type IV collagen from other species. AB769 has not been tested with other extracellular matrix proteins (e.g., laminin, fibronectin).
Dot Blotting, ELISA, Immunocytochemistry, Immunohistochemistry
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
Collagen Type IV Ab-3
Affinity Purified
Human and bovine placental collagen type IV
RUO
Primary
Bovine, Human, Murine
Unconjugated
Goat
200μg
Cell Structure
Polyclonal