Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Collagen V, Biotin, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP126552

Catalog No. NBP126552


Add to cart

Description

Description

Collagen V Polyclonal antibody specifically detects Collagen V in Human, Bovine, Equine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Dot Blot, Immunocytochemistry.
Specifications

Specifications

Collagen V
0.4mg/mL
Western Blot 1:100-1:2000, ELISA 1:1000-1:4000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:20, Immunohistochemistry-Frozen 1:20-1:40, Dot Blot 1:100-1:2000, Immunocytochemistry
Affinity Purified
COL5A1
Goats were hyperimmunized with human type V collagen.
Immunogen affinity purified
RUO
Primary
1289
Bovine, Equine, Human
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Biotin
PBS with Sodium Azide
alpha 1 type V collagen, Collagen 5, collagen alpha-1(V) chain, collagen, type V, alpha 1, Collagen-5, EC 2.7.7.6, EC 6.1.1
Goat
IgG
0.1 mg
Store at 4C. Do not freeze.
Polyclonal
Reacts with conformational determinants on human type V collagen as demonstrated by ELISA. May react with type V collagen from other species. Exhibits <10% cross reactivity with collagen type I, II, III, IV and VI. The antibody has not been tested for reactivity with other ECM proteins (e.g., laminin, fibronectin).
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only