Learn More
Collagen V Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136066
Catalog No. 01-672-249
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reconstitute with 1 mL of distilled water and add preservative if preferred.
Type V collagen is found in tissues containing type I collagen and appears to regulate the assembly of heterotypic fibers composed of both type I and type V collagen. This gene product is closely related to type XI collagen and it is possible that the collagen chains of types V and XI constitute a single collagen type with tissue-specific chain combinations. Mutations in this gene are associated with Ehlers-Danlos syndrome, types I and II.
Specifications
Specifications
Collagen V
Polyclonal
whole serum with no preservative
P20908
RP11-263F14.1
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
ELISA, Immunohistochemistry (Frozen)
Unconjugated
COL5A1
Lyophilized
COL5A1
Purified native collagen type V.
1 mL
4° C
Primary
1289