Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen V Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA136067

Catalog No. 01-672-250


Add to cart

Description

Description

Reconstitute with 1 mL of distilled water and add preservative if preferred.

Type V collagen is found in tissues containing type I collagen and appears to regulate the assembly of heterotypic fibers composed of both type I and type V collagen. This gene product is closely related to type XI collagen and it is possible that the collagen chains of types V and XI constitute a single collagen type with tissue-specific chain combinations. Mutations in this gene are associated with Ehlers-Danlos syndrome, types I and II.
Specifications

Specifications

Collagen V
Polyclonal
Unconjugated
COL5A1
Lyophilized
COL5A1
Purified collagen type V extracted from human skin .
Affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Immunohistochemistry (Frozen)
0.1 mg/mL
Antibody with no preservative
P20908
RP11-263F14.1
Rabbit
IgG
1 mL
4° C
Primary
1289
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit