Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Collagen VIII alpha 1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner 25900002

Catalog No. 25-900-002


Add to cart

Description

Description

Collagen VIII alpha 1 Polyclonal antibody specifically detects Collagen VIII alpha 1 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Collagen VIII alpha 1
Unconjugated
20mM Potassium Phosphate (pH 7.0) and 0.15M NaCl with No Preservative
Affinity Purified
COL8A1
This antibody was made against a protein fragment from the N Terminus Region
Immunogen affinity purified
RUO
Primary
1295
Human
ELISA, Immunocytochemistry, Immunofluorescence
ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:250-1:2000
P27658
alpha-1 polypeptide, chromosome 3 open reading frame 7, collagen, type VIII, alpha 1, smooth muscle cell-expressed and macrophage conditioned medium-induced proteinsmag-64
Rabbit
IgG
0.05 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
This product is specific for Human COL8A1.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only