Learn More
COL23A1 Mouse anti-Human, Clone: 2C9, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA522849
Catalog No. PIMA522849
Add to cart
Description
Description
Peptide Sequence: KGELGLPGAP GIDGEKGPKG QKGDPGEPGP AGLKGEAGEM GLSGLPGADG LKGEKGESAS DSLQESLAQL IVE
COL23A1 is a member of the transmembrane collagens, a subfamily of the nonfibrillar collagens that contain a single pass hydrophobic transmembrane domain (Banyard et al., 2003 [PubMed 12644459]).
Specifications
Specifications
COL23A1
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.4
Q86Y22
COL23A1, collagen alpha-1(XXIII) chain, collagen, type XXIII, alpha 1, DKFZp434K0621, procollagen, type XXIII, alpha 1
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
91522
ELISA, Western Blot
2C9
Unconjugated
COL23A1
Liquid
COL23A1
COL23A1 (NP_775736, 338 a.a. ∽ 410 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human