Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C3 Chicken anti-Human, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA120868

Catalog No. PA120868


Add to cart

Description

Description

PA1-20868 detects C3 in human samples. PA1-20868 has been successfully used in ELISA and Western blot procedures. The PA1-20868 immunogen is human plasma protein. Store at 4°C short term. For long term storage, aliquot and store at -20°C, avoiding freeze/thaw cycles.

The complement factor C3 consists of an alpha and a beta chain. C3 is a central factor in the complement cascade. It is central to the alternative pathway that leads to the C3 convertase C3bBb. The classical mannose binding lectin activation pathway leads to the C3 convertase C4b2a. These convertases cleave C3 resulting in C3a andC3b. Further degradation leads to the formation of the alpha chain products C3d, C3g and C3c. C3 is an acute phase protein that is produced by a wide range of tissues, including renal epithelial cells and hepatocytes.
Specifications

Specifications

Complement C3
Polyclonal
Unconjugated
C3
Liquid
C3
Human plasma protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Western Blot
1.3 mg/mL
PBS with 0.02% sodium azide
P01024
AHUS5, ARMD9, ASP, CPAMD1
Chicken
IgY
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
718
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit