Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C3c Sheep anti-Human, FITC, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Manufacturer:  Invitrogen PA136179

Catalog No. 01-672-128


Add to cart

Description

Description

Collagen IV is a major constituent of the basement membranes along with laminins and enactins. It is composed of alpha 1 IV chain and alpha 2 IV chain in 2:1 ratio. It can form insoluble fibers with high tensile strength. Antibody to collagen IV is useful in detecting the loss of parts of basement membrane in carcinomas.
Specifications

Specifications

Complement C3c
Polyclonal
PBS with 0.09% sodium azide; pH 7.2
P01024
AHUS5, ARMD9, ASP, CPAMD1
Sheep
IgG
1 mL
4° C
Primary
718
Immunohistochemistry (Frozen)
FITC
C3
Liquid
C3
Human C3 purified from serum.
Purified
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit