Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C3c Sheep anti-Human, HRP, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Manufacturer:  Invitrogen PA186651

Catalog No. PA186651


Add to cart

Description

Description

Sheep anti Human C3c antibody detects human complement 3c, a polypeptide fragment which is present in C3.

Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacterial infection.
Specifications

Specifications

Complement C3c
Polyclonal
HRP
C3
Liquid
C3
Native human C3c purified from serum
Protein G
RUO
Antibody
Polyclonal
Human
ELISA, Western Blot
1 mg/mL
PBS with proprietary stabilizer and 0.01% thimerosal; pH 7.2
P01024
AHUS5, ARMD9, ASP, CPAMD1
Sheep
IgG
1 mL
4° C, do not freeze
Primary
718
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit