Learn More
C2 Mouse anti-Human, Clone: 5C3, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen HYB0500402
Catalog No. HYB0500402
Add to cart
Description
Description
HYB 050-04-02 is specific for C2 from human plasma/serum. Crossreactivity to C2 from other species has not been tested.  HYB 050-04-02 has been successfully used in ELISA, and Western blotting procedures. HYB 050-04-02 reacts with human C2 in both native and denatured forms. In Western blotting after SDS-PAGE, HYB 050-04-02 reacts with C2 and a subfraction of C2 believed to be C2b, both under reducing and non-reducing conditions. In ELISA, strong reaction is seen with C2 directly coated onto the microtiter well. The HYB 050-04-02 immunogen is complement component C2 isolated from human plasma. Epitope specificity of HYB 050-04-02 differs from that of HYB 050-05-02 and HYB 050-08-02 as determined by inhibition ELISA. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
Complement component C2 is a component of the classical complement pathway and is involved in activation of C3 and C5. C2 consists of a single polypeptide chain with a molecular mass of 102 kDa.
Specifications
Specifications
C2
Monoclonal
0.90 mg/mL
PBS with 15mM sodium azide; pH 7.4
P06681
C2, C3/C5 convertase, CO2, Complement C2
Mouse
IgG2a, kappa
200 μL
4° C, store in dark
Primary
717
ELISA, Western Blot
5C3
Unconjugated
C2
Liquid
C2
Complement component C2 isolated from human plasma.
Protein A
RUO
Antibody
Monoclonal
Human