Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement C3 Mouse anti-Human, Clone: 2D8, Invitrogen
Mouse Monoclonal Antibody
Manufacturer: Invitrogen HYB0050102
Description
HYB 006-02-02 detects human Glucogon-like protein from human samples. Cross-reactivity with hGLP-2 from other species has not been determined. HYB 006-02-02 has been successfully used in ELISA procedures. HYB 006-02-02 reacts with hGLP-2 in sandwich ELIS
C3 is a key component of the complement system since classical and alternative activation pathways merge at the C3 activation step when C3 is split into C3a and C3b. The molecular mass of C3 is 185 kDa and it consists of two chains (110 kDa and 75 kDa) held together by disulfide bonds.Specifications
Complement C3 | |
Monoclonal | |
0.92 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P01024 | |
C3, Acylation Stimulating Protein Cleavage Product | |
Mouse | |
IgG1, kappa | |
200 μL | |
4° C, store in dark | |
Primary | |
718 |
ELISA, Western Blot | |
2D8 | |
Unconjugated | |
C3 | |
Liquid | |
C3 | |
C3 isolated from human plasma. | |
Protein A | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title