Learn More
Connexin 37 Sheep anti-Mouse, Rat, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen PA118407
Catalog No. 01-670-657
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA1-18407 detects Connexin-37 from mouse and rat samples. PA1-18407 has been successfully used in immunohistochemistry. The PA1-18407 immunogen is a synthetic peptide corresponding to residues Y(265) L P M G E G P S S P P C P T Y(280) of rat Connexin-37, conjugated to diphtheria toxoid. Reconstitute with 100 μL of distilled water.
Gap junction protein that mediates cell-cell communication; may mediate hyperexcitability following nerve injury.
Specifications
Specifications
Connexin 37
Polyclonal
whole serum with no preservative
P28235, Q03190
Gap junction alpha-4 protein, Cx37, Gja4, Cxn-37
Sheep
IgG
RUO
Antibody
Polyclonal
Mouse, Rat
Immunohistochemistry, Western Blot
Unconjugated
Gja4
Lyophilized
GJA4
Synthetic peptide corresponding to residues Y(265) L P M G E G P S S P P C P T Y(280) of rat Connexin-37.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
14612, 25655