Learn More
MilliporeSigma™ anti-Connexin 43, C-terminus, cytosolic ; Polyclonal; ™ (Chemicon™) Non-distribution product as customer accommodation.
Manufacturer:  MilliporeSigma™ AB1728/DEL
Catalog No. 50-173-241
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Beyer & Steinberg 1991; Nishi et al. 1991; Hennemann et al. 1992
ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
Connexin 43, C-terminus, cytosolic
Affinity Purified
Anti-Connexin 43 is made against a 23 amino acid C-terminal peptide sequence within the cytoplasmic domain of mouse Cx43. Peptide was coupled to KLH (carrier protein).
RUO
Primary
Human, Murine, Rat
Unconjugated
Rabbit
50μg
Cell Structure
Polyclonal