Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Connexin 43/GJA1 APC-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems FAB7737A
This item is not returnable.
View return policy
Description
Connexin 43/GJA1 Monoclonal specifically detects Connexin 43/GJA1 in Human samples. It is validated for Flow Cytometry.Specifications
Connexin 43/GJA1 | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
connexin 43, connexin-43, Cx43, CX43gap junction protein, alpha-like, DFNB38, Gap junction 43 kDa heart protein, gap junction alpha-1 protein, gap junction protein, alpha 1, 43kDa, gap junction protein, alpha 1, 43kDa (connexin 43), GJA1, GJAL, HSS, ODD, ODDD, ODOD, SDTY3 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2697 | |
Human | |
Purified |
Flow Cytometry | |
578618 | |
Flow Cytometry 10 uL/10^6 cells | |
P17302 | |
GJA1 | |
Mouse myeloma cell line NS0 transfected with human Connexin 43/GJA1 Accession # P17302 | |
100 Tests | |
Primary | |
Detects human Connexin 43/GJA1 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction