Learn More
Contactin 6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSC00023W
Catalog No. PIOSC00023W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water.
Specifications
Specifications
Contactin 6
Polyclonal
whole serum with no preservative
Q9UQ52, Q9JMB8, P97528
Neural recognition molecule NB-3, NB3, contactin 6, CNTN6
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry, Western Blot
Unconjugated
CNTN6
Lyophilized
CNTN6
A synthetic peptide from aa region 930-980 of human Contactin 6 conjugated to an immunogenic carrier protein was used as the antigen
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
27255, 27256, 53870