Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COPS3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA515026

Catalog No. PIPA515026

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

COPS3 possesses kinase activity that phosphorylates regulators involved in signal transduction. This protein phosphorylates I kappa-Balpha, p105, and c-Jun. It acts as a docking site for complex-mediated phosphorylation.
Specifications

Specifications

COPS3
Polyclonal
PBS with 0.09% sodium azide
Q9UNS2
COP9 signalosome complex subunit 3; JAB1-containing signalosome subunit 3; CSN3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8533
Western Blot
Unconjugated
COPS3
Liquid
COPS3
KLH conjugated synthetic peptide between 98-130 amino acids from the central region of human COPS3
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit