Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COQ5 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526327

Catalog No. PIPA526327


Add to cart

Description

Description

This antibody is predicted to react with bovine and rat based on sequence homology.

Specifications

Specifications

COQ5
Polyclonal
Unconjugated
COQ5
Liquid
COQ5
KLH conjugated synthetic peptide between 213-241 amino acids from the central region of human COQ5
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
0.37 mg/mL
PBS with 0.09% sodium azide
Q5HYK3, Q9CXI0
2-methoxy-6-polyprenyl-1,4-benzoquinol methylase, mitochondrial, ubiquinone biosynthesis methyltransferase COQ5, mitochondrial
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
52064, 84274
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit