Learn More
COQ7 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA525774
Catalog No. PIPA525774
Add to cart
Description
Description
Studies have shown that murine HSP90 exists as two forms, HSP84 and HSP86, coded by related but separate genes, with 86% amino acid sequence conservation. These forms are analogous to the two forms of human HSP90, HSP89 alpha and HSP89 beta. In an unstressed mouse fibroblast, the basal level of HSP84 is found to be double that of HSP86. However, after heat shock, HSP86 shows a greater increase. Studies also suggest that upon cellular differentiation, the level of HSP86, but not HSP84, decreases. HSP84 and HSP86, which may be subject to estrogenic regulation, have been found as components of the non-DNA binding form of mouse glucocorticoid receptor, but dissociated from the transformed DNA-binding form.
Specifications
Specifications
COQ7
Polyclonal
Unconjugated
COQ7
Liquid
COQ7
KLH conjugated synthetic peptide between 162-191 amino acids from the C-terminal region of human COQ7
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q99807
CAT5, CLK-1, CLK1, COQ7 coenzyme Q, 7 homolog ubiquinone, coenzyme Q biosynthesis protein 7 homolog, placental protein KG-20, timing protein clk-1 homolog, ubiquinone biosynthesis protein COQ7 homolog
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10229