Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CORIN Rat anti-Human, Clone: 231443, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA523899

Catalog No. PIMA523899


Add to cart

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

Specifications

Specifications

Corin
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q9Y5Q5
ATC2, Pro-ANP-Converting Enzyme, atrial natriuteric peptide-converting enzyme, Corin, corin, serine peptidase, EC 3.4.21, EC 3.4.21.-, heart specific serine proteinase, Heart-specific serine proteinase ATC2, MGC119742, pro-ANP-convertase, Pro-ANP-converting enzyme, PRSC, serine protease, Transmembrane protease serine 10
Rat
IgG2A
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
10699
Immunocytochemistry, Immunofluorescence, Western Blot
231443
Unconjugated
CORIN
Lyophilized
CORIN
Mouse myeloma cell line NS0-derived recombit human Corin Ser67-Asn1042
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit