Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coronin 7 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524255

Catalog No. PIPA524255


Add to cart

Description

Description

Coronins, such as CORO7, constitute an evolutionarilyconserved family of WD-repeat actin-binding proteins. CORO7 plays arole in Golgi complex morphology and function.
Specifications

Specifications

Coronin 7
Polyclonal
PBS with 0.09% sodium azide
P57737
0610011B16Rik, CRN7, POD1, 70 kDa WD repeat tumor rejection antigen homolog, coronin-7
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79585
Immunohistochemistry, Western Blot
Unconjugated
CORO7
Liquid
CORO7
KLH conjugated synthetic peptide between 173-202 amino acids from the N-terminal region of human CORO7
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit