Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coronin 1A Chicken anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA110007

Catalog No. 01-670-264


Add to cart

Description

Description

Highly conserved coronin family of proteins/actin regulators from WD-repeat superfamily were originally identified in Dictyostelium, wherein, coronin deletions were observed to affect actin-dependent processes, such as phagocytosis, cell motility, and cytokinesis. Coronin-1A (Coro1A) belongs to Type I coronins, almost exclusively expressed in hematopoietic lineages, and plays a key role during T lymphocyte activation, migration, survival, and calcium signaling. Moreover, Coronin-1A has been documented to be dispensable for actin-dependent functions in macrophages. Coro1a-deficient mice have reduced peripheral T cells due to both an impaired survival of naive cells and migrational defects. Both CD4+ and CD8+ T cell subsets are decreased in the periphery of Coro1a-/- mice while thymic cellularity and subpopulations are similar to those observed in wild-type mice. A nonsense mutation in Coro1a has been shown to protect against systemic lupus in a mouse model. Moreover, it has shown that Coro1a is mutated in a thymic egress-deficient mouse strain, but also in a patient with severe combined immunodeficiency (SCID).
Specifications

Specifications

Coronin 1a
Polyclonal
PBS with 0.02% sodium azide
P31146, O89053, Q91ZN1
CLABP, CLIPINA, FLJ41407, HCORO1, MGC117380, TACO, p57
Chicken
IgY
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
11151, 12721, 155151
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
CORO1A
Liquid
CORO1A
Purified, recombit human coronin 1a.
PEG precipitation
RUO
Antibody
Polyclonal
Human, Mouse, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit