Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cortactin Mouse, Unlabeled, Clone: 30, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610050

 View more versions of this product

Catalog No. BDB610050


Add to cart

Description

Description

Cortactin is a protein which is represented by two structurally related isoforms of 80 kDa and 85 kDa. Sequence analysis of this protein reveals the presence of a carboxy-terminal SH3 domain, as well as six internal tandem repeats of a 37 amino acid sequence. It is able to bind to F-actin via this N-terminal repeat region, and colocalizes with actin in the cell. Cortactin was originally described as a protein that is tyrosine-phosphorylated in cells transformed by the src oncogene, in which it localizes to rosettes or podosomes. It has since been shown that other stimuli, such as bacterial invasion, treatment with growth factors, and integrin-mediated cell adhesion, are also capable of inducing phosphorylation of cortactin. Some additional evidence indicates that cortactin may actually be a substrate of syk, with src acting as an upstream signaling molecule.
Specifications

Specifications

Cortactin
30
Unconjugated
Affinity Purified
Chicken Cortactin
150μg
Cell Biology
Primary
Canine, Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.