Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cortactin (p80/85) Mouse anti-Avian, Bovine, Human, Hamster, Mouse, Rat, Clone: 4F11, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05180
Description
Anti-Cortactin (p80/85) Antibody, clone 4F11 is an antibody against Cortactin (p80/85), and has been published and validated for use in immunocytochemistry, immunohistochemistry, immunoprecipitation and western blotting.Specifications
Cortactin (p80/85) | |
Monoclonal | |
Unconjugated | |
NM_005231.2; NM_138565.1 | |
Mouse | |
Protein G purfied | |
Primary | |
−20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
4F11 | |
Purified immunoglobulin in 10mM PBS, pH 7.4 with 0.05% sodium azide. | |
CTTN, EMS1, Amplaxin, FLJ34459, 1110020L01Rik | |
Mixture of affinity-purified tyrosine phosphoproteins from chick embryo fibroblasts expressing activated pp60src | |
200 μg | |
Avian, Bovine, Hamster, Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only