Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRF, Polyclonal, Invitrogen™

Catalog No. 01673319
Change view
Click to view available options
Quantity:
7 mL
Catalog No. Quantity
01-673-319 7 mL
1 options

Catalog No. 01-673-319

Supplier: Thermo Scientific PA137498

Rabbit Polyclonal Antibody

Corticotropin Releasing Factor (CRF), a 41 amino acids neuropeptide, belongs to a family of structurally related peptides including urocortin, sauvagine and urotensin I. CRF is critical in the regulation of the pituitary-adrenal gland axis, and in endocrine, autonomic and behavioral responses to stress. CRF is widely distributed in the brain and peripheral nervous system with the highest levels in the hypothalamic paraventricular nucleus.

This antibody is predicted to react with chicken, canine, amphibian, mouse, porcine and rat based on sequence homology.Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is brain or placenta tissue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRF
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Gene CRH
Gene Accession No. P06850
Gene Alias Corticotropin Releasing Factor
Gene Symbols CRH
Host Species Rabbit
Immunogen Synthetic peptide from C-terminus of human CRF protein
Purification Method Antigen affinity chromatography
Quantity 7 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1392
Target Species Human
Content And Storage Store at 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.