Learn More
Corynebacterium diphtheriae Toxin Mouse anti-Bacteria, Clone: 11D9, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen HYB1230902
Catalog No. 01-670-120
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
HYB 123-09-02 detects Corynebacterium diphtheriae toxoid in bacteria samples. HYB 123-09-02 has been successfully used in ELISA and Western blot procedures. Where a band (∽63 kDa) is detected corresponding to the toxin proenzyme. HYB 123-09-02 immunogen is Diphtheria toxoid prepared from C.diphtheria subsp. Intermedius Park-Williams no. 8 by formaldehyde inactivation. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
Diphtheria toxin (DT) is secreted by certain strains of Corynebacterium diphtheriae and catalyzes the ADP-ribosylation of eukaryotic aminoacyl-transferase II (EF-2) using NAD as a substrate. This reaction forms the basis for its toxicity toward eukaryotic organisms. Diphtheria toxin is synthesized and excreted as a proenzyme, composed of a single polypeptide chain having a molecular weight of approximately 63 kDa. Two covalent alterations in structure are necessary for expression of its enzymatic activity. First, mild proteolysis results in the formation of "nicked toxin," which is enzymatically inactive and consists of two major fragments, A and B, linked by a disulfide bond. Reduction of the nicked toxin with thiols releases the N-terminal A fragment (∽24 kDa) which is enzymatically active. The C-terminal B fragment (∽39 kDa) has no apparent enzymatic activity, but is required for toxicity. Evidence suggests that the B fragment is responsible for recognizing and binding the toxin to cell surface receptors.
Specifications
Specifications
Corynebacterium diphtheriae Toxin
Monoclonal
1.06 mg/mL
PBS with 15mM sodium azide; pH 7.4
C. diphtheriae; Diphtheria
Diphtheria toxoid (formaldehyde inactivated Diphtheria toxin).
Protein A
RUO
Antibody
Monoclonal
ELISA, Western Blot
11D9
Unconjugated
Liquid
Mouse
IgG1, kappa
200 μL
4° C, store in dark
Primary
Bacteria