Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotin Anti-Cox-2 Mouse, Biotin, Clone: 33, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610205

Catalog No. BDB610205


Add to cart

Description

Description

Cyclooxygenase (Cox) is also known as prostaglandin H synthase or PGH synthase (E.C. 1.14.99.1). It catalyzes the conversion of arachidonate to prostaglandin H2 (PGH2), the precursor of PGE2, PGF2α, PGD2, prostacyclin, and thromboxane A2. Cox actually has two different enzymatic activities: a cyclooxygenase that mediates the formation of PGG2 from oxygen andarachidonate and a hydroperoxidase that catalyzes a reduction of PGG2 yielding PGH2. Two Cox genes, Cox-1 and Cox-2, have been isolated in several species. A 4kb mRNA encodes the 604 amino acid Cox-2 protein. The two human Cox isoenzymes are 61% identical in amino acid composition with the active sites being highly conserved. Cox-2 mRNA and protein levels are induced by serum, lipopolysaccharides, growth factors, human chorionic gonadotropin and phorboltesters in various mammalian cell types. It has been shown that interleukin-1α (IL-1α) induces increased levels of Cox-2 mRNA and protein in human endothelial cells. The sustained increase in Cox-2 is apparently due (at least in part) to IL-1α increasing the stability of Cox-2 mRNA. This type of regulatory mechanism may play an important role in chronic inflammatory conditions.

Specifications

Specifications

Biotin Anti-Cox-2
33
Biotin
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
PGHS-2, Cyclooxygenase-2
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Chicken, Human, Murine
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit