Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA137504

Catalog No. 01-673-323


Add to cart

Description

Description

This antibody is predicted to react with bovine, mouse and rabbit based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is colon, lung or breast carcinoma.

COX2 (Cyclooxygenase 2) is an inducible enzyme. It is involved in the response of cells to growth factors, tumor promoters, and cytokines that induce its expression. Given its role in synthesizing prostaglandins, COX2 is therefore of interest in studying immune response regulation. COX2 is induced by a wide variety of stimuli and was initially identified as immediate early growth response gene. In addition, COX2 expression markedly increased in 85-90% of human colorectal adenocarcinoma whereas COX1 levels remain unchanged.
Specifications

Specifications

COX2
Polyclonal
TBS with 1% BSA and 0.099% sodium azide
P35355, P35354, Q05769
Cyclooxygenase 2
Rabbit
IgG
7 mL
4° C, do not freeze
Primary
5743, 19225, 29527
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PTGS2
Liquid
PTGS2
Synthetic peptide derived from C-terminus of rat COX-2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit