Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX5A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526161

Catalog No. PIPA526161

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with rabbit based on sequence homology.

This is the heme A-containing chain of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.
Specifications

Specifications

COX5A
Polyclonal
PBS with 0.09% sodium azide
P20674
COX, COX-VA, VA, cytochrome c oxidase polypeptide Va, cytochrome c oxidase polypeptide, mitochondrial, cytochrome c oxidase subunit 5A, mitochondrial, mitochondrial cytochrome c oxidase subunit Va
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9377
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
COX5A
Liquid
COX5A
KLH conjugated synthetic peptide between 46-73 amino acids from the central region of human COX5A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit