Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-COX6A2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP131112

Catalog No. NBP131112


Add to cart

Description

Description

COX6A2 Polyclonal antibody specifically detects COX6A2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

COX6A2
0.9mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
COX6A2
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human COX6A2. The exact sequence is proprietary.
11 kDa
0.1 ml
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Predicted cross-reactivity based on sequence identity: Rat (86%). Not yet tested in any other species.
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
COX VIa-M, COX6A, COX6AH, COXVIAH, Cytochrome c oxidase polypeptide VIa-heart, cytochrome c oxidase subunit 6A2, mitochondrial, cytochrome c oxidase subunit VIa polypeptide 2, Cytochrome c oxidase subunit VIA-muscle
Rabbit
IgG
Antigen Affinity-purified
RUO
Primary
1339
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only