Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cytochrome c oxidase subunit Vib polypeptide 1 (ubiquitous), Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human COX6B1 protein.

Manufacturer:  Abnova Corporation H00001340B01

Catalog No. 89-005-163


Add to cart

Description

Description

Cytochrome c oxidase (COX), the terminal enzyme of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. It is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may be involved in the regulation and assembly of the complex. This nuclear gene encodes subunit VIb. Three pseudogenes COX6BP-1, COX6BP-2 and COX6BP-3 have been found on chromosomes 7, 17 and 22q13.1-13.2, respectively. [provided by RefSeq

Sequence: MAEDMETKIKNYKTAPFDSRFPNQNQTRNCWQNYLDFHRCQKAMTAKGGDISVCEWYQRVYQSLCPTSWVTDWDEQRAEGTFPGKI
Specifications

Specifications

cytochrome c oxidase subunit Vib polypeptide 1 (ubiquitous)
Unconjugated
No additive
NM_001863.3
COX6B|COXG
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
1340
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human COX6B1 protein.
COX6B1
Serum
COX6B1
COX6B1 (NP_001854.1, 1 a.a. ~ 86 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit