Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPEB4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525538

Catalog No. PIPA525538


Add to cart

Description

Description

There are 3 named isoforms produced by alternative splicing.
Specifications

Specifications

CPEB4
Polyclonal
Unconjugated
CPEB4
Liquid
CPEB4
KLH conjugated synthetic peptide between 109-137 amino acids from the N-terminal region of human CPEB4
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q17RY0, Q7TN98
CPE-BP4, CPE-binding protein 4, cytoplasmic polyadenylation element-binding protein 4, hCPEB-4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
67579, 80315
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit