Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPE1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527668

Catalog No. PIPA527668


Add to cart

Description

Description

PA5-27668 targets Cpn10 in WB applications and shows reactivity with Human and mouse samples. The PA5-27668 immunogen is recombit protein fragment corresponding to a region within amino acids 1 and 91 of Human Cpn10. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a major heat shock protein which functions as a chaperonin. Its structure consists of a heptameric ring which binds to another heat shock protein in order to form a symmetric, functional heterodimer which enhances protein folding in an ATP-dependent manner. This gene and its co-chaperonin, HSPD1, are arranged in a head-to-head orientation on chromosome 2.
Specifications

Specifications

HSPE1
Polyclonal
Unconjugated
HSPE1
Liquid
HSPE1
Recombit protein fragment corresponding to a region within amino acids 1 and 91 of Human Cpn10
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
1 mg/mL
PBS with 40% glycerol and 0.01% thimerosal; pH 7
P61604, Q64433
CPN10; EPF; GROES; HSP10; 10 kDa chaperonin; 10 kDa heat shock protein; mitochondrial; chaperonin 10; early-pregcy factor; heat shock 10kD protein 1 (chaperonin 10); CH10
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
15528, 3336
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit