Learn More
CPP Binding Protein Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA3106
Catalog No. PA3106
Add to cart
Description
Description
PA3-106 detects the CPP-binding protein. PA3-106 has been used successfully in Western blot, ELISA, immunoprecipitation, immunohistochemistry, and immunocytochemistry procedures. In Western blot analysis of rat brain synaptic membranes this antibody detects ~80 kDa and ~60 kDa proteins representing CPP-binding protein. The ~60 kDa protein is likely a breakdown product of the ~80 kDa CPP-binding protein. The PA3-106 immunogen is a synthetic peptide corresponding to residues (68)K K R K K K S S E T F L S E L A E I(85) C of rat CPP-binding protein.
CPP is a potent antagonist acting selectively at NMDA type excitatory amino acid receptors. This compound is more potent than previously reported NMDA antagonists in depressing mammalian spinal neuronal responses.
Specifications
Specifications
CPP Binding Protein
Polyclonal
whole serum with 0.05% sodium azide
Rabbit
IgG
RUO
Antibody
Polyclonal
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
Liquid
Synthetic peptide corresponding to residues K(68) K R K K K S S E T F L S E L A E I(85) C of rat CPP-binding protein.
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
Human, Mouse, Rat