Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CPT2, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP146137

Catalog No. NBP146137


Add to cart

Description

Description

CPT2 Polyclonal antibody specifically detects CPT2 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications

Specifications

CPT2
0.5 mg/mL
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:4000
NP_000089.1
carnitine O-palmitoyltransferase 2, mitochondrial, carnitine palmitoyltransferase 2, Carnitine palmitoyltransferase IIEC 2.3.1.21, CPT II, CPT1, CPTASE
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody recognizes and epitope within the internal region.
ELISA, Western Blot
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
CPT2
Peptide with sequence C-TDSTVTVQKLNFE corresponding to internal region according to NP_000089.1.
Immunogen affinity purified
RUO
Primary
1376
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only