Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HID1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525132

Catalog No. PIPA525132

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse based on sequence homology.

CQ028 may play an important role in the development of cancers in a broad range of tissues.
Specifications

Specifications

HID1
Polyclonal
PBS with 0.09% sodium azide
Q8IV36, Q8R1F6
DMC1, UPF0663 transmembrane protein C17orf28, down-regulated in multiple cancers 1, downregulated in multiple cancer 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
217310, 283987
Western Blot
Unconjugated
HID1
Liquid
HID1
KLH conjugated synthetic peptide between 567-595 amino acids from the C-terminal region of human CQ028
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit