Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C18orf25 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524485

Catalog No. PIPA524485


Add to cart

Description

Description

The exact function of C18orf25 remains unknown. There are two named isoforms.
Specifications

Specifications

C18orf25
Polyclonal
PBS with 0.09% sodium azide
Q96B23, Q8BH50
ARKL1, ARKadia-like 1, uncharacterized protein C18orf25
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
147339, 212163
Immunohistochemistry, Western Blot
Unconjugated
C18orf25
Liquid
8030462N17Rik, C18orf25
KLH conjugated synthetic peptide between 163-191 amino acids from the central region of human CR025
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit