Learn More
CRACC (CD319) Mouse anti-Human, Alexa Fluor 647, Clone: 235614, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 564338
Catalog No. BDB564338
Add to cart
Description
Description
The 235614 monoclonal antibody specifically binds to CRACC (CD2-like receptor activating cytotoxic cells) which is also known as CD319, CS1 (CD2 subset 1), SLAMF7, 19A, and 19A24. CRACC is a type I transmembrane glycoprotein that belongs to the CD2 family of receptors within the Ig superfamily. CRACC is expressed on most NK cells and subsets of CD8+ T cells, CD4+ T cells, B cells, macrophages, and dendritic cells. CRACC reportedly plays roles in the activation, proliferation and effector functions of T cells, NK cells, and other leucocytes.
Specifications
Specifications
CRACC (CD319)
235614
CRACC; SLAMF7; 19A; 19A24 protein; CS1
Affinity Purified
Human CRACC/SLAMF7
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Alexa Fluor 647
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG2a, κ
RUO
Primary
Human