Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATF6-beta Rabbit anti-Hamster, Human, Mouse, Polyclonal, Invitrogen
Rabbit Polyclonal Antibody
Manufacturer: Invitrogen PA525025
Description
CREBL1 is a transcription factor in the unfolded protein response (UPR) pathway during ER stress. Either as a homodimer or as a heterodimer with ATF6-alpha, the encoded protein binds to the ER stress response element, interacting with nuclear transcription factor Y to activate UPR target genes. The protein is normally found in the membrane of the endoplasmic reticulum; however, under ER stress, the N-terminal cytoplasmic domain is cleaved from the rest of the protein and translocates to the nucleus.Specifications
ATF6-beta | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q99941, O35451 | |
CREB-RP, CREBL1, G13, ATF6-beta, Creb-related protein, cAMP response element-binding protein-related protein, cAMP responsive element binding protein-like 1, cAMP-dependent transcription factor ATF-6 beta, cAMP-responsive element-binding protein-like 1, cyclic AMP-dependent transcri | |
Rabbit | |
IgG | |
400 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
100767112, 12915, 1388 |
Western Blot | |
Unconjugated | |
ATF6B | |
Liquid | |
ATF6B | |
KLH conjugated synthetic peptide between 560-589 amino acids from the C-terminal region of human CREBL1 | |
Antigen affinity chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Hamster, Human, Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title