Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRELD2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526192

Catalog No. PIPA526192


Add to cart

Description

Description

CRELD2 may regulate transport of alpha4-beta2 neuronal acetylcholine receptor.
Specifications

Specifications

CRELD2
Polyclonal
PBS with 0.09% sodium azide
Q6UXH1
cysteine-rich with EGF-like domain protein 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79174
Flow Cytometry, Western Blot
Unconjugated
CRELD2
Liquid
CRELD2
KLH conjugated synthetic peptide between 325-353 amino acids from the C-terminal region of human CRELD2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit