Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CREM Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526131

Catalog No. PIPA526131


Add to cart

Description

Description

CREM is a bZIP transcription factor that binds to the cAMP responsive element found in many viral and cellular promoters. It is an important component of cAMP-mediated signal transduction during the spermatogenetic cycle, as well as other complex processes.
Specifications

Specifications

CREM
Polyclonal
PBS with 0.09% sodium azide
Q03060
CREM-2, ICER, hCREM-2, CREM 2alpha-b protein, CREM 2beta-a protein, cAMP response element modulator, cAMP-responsive element modulator, inducible cAMP early repressor ICER
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1390
Flow Cytometry, Western Blot
Unconjugated
CREM
Liquid
CREM
KLH conjugated synthetic peptide between 287-314 amino acids from the C-terminal region of human CREM
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit