Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRHR1 Monoclonal Antibody (343919), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA524079
Description
This antibody detects human CRHR-1 transfectants but not irrelevant transfectants. Reconstitute at 0.5 mg/mL in sterile PBS.
CRHR1 Monoclonal antibody specifically detects CRHR1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
CRHR1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P34998 | |
CRHR1 | |
NS0 mouse myeloma cell line transfected with human CRHR-1 Ser24-Val415 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry | |
343919 | |
Unconjugated | |
CRHR1 | |
corticotrophin releasing hormone receptor 1; corticotropin releasing hormone 1; corticotropin releasing hormone receptor 1; corticotropin-releasing factor receptor 1; corticotropin-releasing factor type 1 receptor; corticotropin-releasing hormone receptor 1; CRF 1 receptor; Crf receptor 1; Crf type 1; CRF1; Crf1 receptor; CRFR; CRF-R; CRFR1; CRF-R1; CRF-R-1; CRFR-1; CRF-R1alpha; CRH receptor 1; CRH receptor subtype-1 isoform-3; Crhr; CRH-R 1; CRHR1; CRH-R1; CRH-R-1; CRHR1f; CRH-R1h; CRHR1L; seven transmembrane helix receptor | |
Mouse | |
Protein A/G | |
RUO | |
1394 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'